Learn More
Research Products International Corp Foil Cover-All, 2 Inch Top Diameter, 5.5 Inch Bottom Diameter, 1.75 Inch Height, 252 per Case
SDP
Catalog No. 50998400
Encompass
Foil Cover-All provides dust free and clean storage of tube, bottle and flask contents. Pre-formed foil cover quickly protects a variety of laboratory containers using only one hand. Keep containers clean, covered, dust and contamination free. When used during autoclaving Cover-Alls provide additional container sterility and ensure container integrity until future use.
WARNING  This product can expose you to chemicals including Chromium, which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-998-400 Supplier Research Products International Corp Supplier No. 246413
Foil Cover-All provides dust free and clean storage of tube, bottle and flask contents. Pre-formed foil cover quickly protects a variety of laboratory containers using only one hand. Keep containers clean, covered, dust and contamination free. When used during autoclaving Cover-Alls provide additional container sterility and ensure container integrity until future use.
WARNING  This product can expose you to chemicals including Chromium, which is known to the State of California to cause cancer or birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.