Learn More
Sigma Aldrich Fine Chemicals Biosciences foil/poly bag size 18 in.
Catalog No. 502899995
Encompass
Heat sealable. Useful for storing air light or moisture-sensitive reagents odoriferous products and for temporarily containing leaking bottles.Recommended heat seal conditions 350f (178C) 40 psi 1 sec.
Catalog No. 50-289-9995 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z1834151PAK
Heat sealable. Useful for storing air light or moisture-sensitive reagents odoriferous products and for temporarily containing leaking bottles.Recommended heat seal conditions 350f (178C) 40 psi 1 sec.