Learn More
Sigma Aldrich Fine Chemicals Biosciences foil/poly bag size 4 1/2
Catalog No. 502901862
Encompass
Heat sealable. Useful for storing air light or moisture-sensitive reagents odoriferous products and for temporarily containing leaking bottles.Recommended heat seal conditions 350f (178C) 40 psi 1 sec.
Catalog No. 50-290-1862 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z1833931PAK
Heat sealable. Useful for storing air light or moisture-sensitive reagents odoriferous products and for temporarily containing leaking bottles.Recommended heat seal conditions 350f (178C) 40 psi 1 sec.