Learn More
Bellco Glass, Inc. Foil Sealing Film reduces sample evaporation can be pierced easily with pipet tips and automation and is heat and cold resistant.Specifications Recommended for use between 80¿C and 120¿C Case Quantity 100
SDP
Catalog No. 502329896
Encompass_Preferred
Foil Sealing Film reduces sample evaporation can be pierced easily with pipet tips and automation and is heat and cold resistant.Specifications Recommended for use between 80¿C and 120¿C Case Quantity 100
Catalog No. 50-232-9896 Supplier Bellco Glass, Inc. Supplier No. 229133000
Foil Sealing Film reduces sample evaporation can be pierced easily with pipet tips and automation and is heat and cold resistant.Specifications Recommended for use between 80¿C and 120¿C Case Quantity 100