Learn More
Bellco Glass, Inc. Foil Sealing Film reduces sample evaporation can be pierced easily with pipet tips and automation and is heat and cold resistant.Specifications Recommended for use between 80¿C and 120¿C Case Quantity 50nbsp
SDP
Catalog No. 502329842
Encompass_Preferred
Foil Sealing Film reduces sample evaporation can be pierced easily with pipet tips and automation and is heat and cold resistant.Specifications Recommended for use between 80¿C and 120¿C Case Quantity 50nbsp
Catalog No. 50-232-9842 Supplier Bellco Glass, Inc. Supplier No. 229132000
Foil Sealing Film reduces sample evaporation can be pierced easily with pipet tips and automation and is heat and cold resistant.Specifications Recommended for use between 80¿C and 120¿C Case Quantity 50nbsp