Learn More
Thomas Scientific Pierce Heat Seal; pierceable heat sealing foil, 100 sheets (125 x 78 mm)
SDP
Catalog No. NC1692369
Encompass
Heat sealing offers a 100% effective method of plate sealing, for complete seal integrity, as well as being quick and cost effective. Pierce Heat Seal is compatible with polypropylene and polystyrene plates. This seal demonstrates good solvent resistance and can be used for low temperature and room temperature compound storage in DMSO and organic solvents.
Catalog No. NC1692369 Supplier Thomas Scientific Supplier No. 1149J10
Heat sealing offers a 100% effective method of plate sealing, for complete seal integrity, as well as being quick and cost effective. Pierce Heat Seal is compatible with polypropylene and polystyrene plates. This seal demonstrates good solvent resistance and can be used for low temperature and room temperature compound storage in DMSO and organic solvents.