Learn More
Grainger Foil Tape 1 in x 5 yd Aluminum
Catalog No. 502749535
Encompass_Preferred
The 3M Foil Tape 15D119 is a 2 inch wide, 60 yard long aluminum foil tape with an acrylic adhesive. It has a temperature resistance range of -20F to 250F and is designed for HVAC sealing and joining applications. This UL listed tape provides a moisture barrier and resists UV light. It bonds well to sheet metal, glass, wood, plastic, and painted surfaces. The acrylic adhesive adheres quickly and forms a long-lasting bond. This tape can be used for ductwork, insulation facing, and foil vapor barrier fabrication. It provides reliable performance across a wide temperature range. The foil backing is flexible and conforms well to irregular surfaces.
Catalog No. 50-274-9535 Supplier Grainger Supplier No. 15D119
The 3M Foil Tape 15D119 is a 2 inch wide, 60 yard long aluminum foil tape with an acrylic adhesive. It has a temperature resistance range of -20F to 250F and is designed for HVAC sealing and joining applications. This UL listed tape provides a moisture barrier and resists UV light. It bonds well to sheet metal, glass, wood, plastic, and painted surfaces. The acrylic adhesive adheres quickly and forms a long-lasting bond. This tape can be used for ductwork, insulation facing, and foil vapor barrier fabrication. It provides reliable performance across a wide temperature range. The foil backing is flexible and conforms well to irregular surfaces.