Learn More
Grainger Foil Tape 1 in x 5 yd Aluminum
Catalog No. 502750649
Encompass_Preferred
The 3M 15D114 Foil Tape is a 2" wide, 60 yard long aluminum foil tape with acrylic adhesive. It has a temperature resistance from -20F to 250F and is used for HVAC sealing, joining sheet metal, and moisture sealing. This UL listed tape can be applied to metal, glass, wood, plastic, and masonry surfaces. It provides a strong bond and conforms well to irregular surfaces. The acrylic adhesive resists UV degradation.
Catalog No. 50-275-0649 Supplier Grainger Supplier No. 15D114
The 3M 15D114 Foil Tape is a 2" wide, 60 yard long aluminum foil tape with acrylic adhesive. It has a temperature resistance from -20F to 250F and is used for HVAC sealing, joining sheet metal, and moisture sealing. This UL listed tape can be applied to metal, glass, wood, plastic, and masonry surfaces. It provides a strong bond and conforms well to irregular surfaces. The acrylic adhesive resists UV degradation.