Learn More
Grainger TapeCase Reinforced Glass Foil Tape, Silver, 1 in. x 5 yd., 1/Pk.
Catalog No. 502755864
Encompass_Preferred
This glass foil tape is designed for high-performance applications requiring thermal resistance and durability. It features a silicone adhesive applied to a reinforced foil backing, making it suitable for demanding environments in assembly, manufacturing, and maintenance tasks.
  • Features a silicone adhesive with an adhesion strength of 40 oz./in.
  • Rated for performance in temperatures from -60°F to +600°F.
  • Constructed with a reinforced foil backing for added strength.
  • Meets MIL-T-83284, Chrysler, Delphi, and Ford specifications.
  • Suitable for both indoor and outdoor use.
  • Continuous roll format with a thickness of 4 mil.
Catalog No. 50-275-5864 Supplier Grainger Supplier No. 15C646
This glass foil tape is designed for high-performance applications requiring thermal resistance and durability. It features a silicone adhesive applied to a reinforced foil backing, making it suitable for demanding environments in assembly, manufacturing, and maintenance tasks.
  • Features a silicone adhesive with an adhesion strength of 40 oz./in.
  • Rated for performance in temperatures from -60°F to +600°F.
  • Constructed with a reinforced foil backing for added strength.
  • Meets MIL-T-83284, Chrysler, Delphi, and Ford specifications.
  • Suitable for both indoor and outdoor use.
  • Continuous roll format with a thickness of 4 mil.