Learn More
Grainger 3M Damping Foil Tape, Aluminum, Silver, 1 in. x 5 yd.
Catalog No. 502757057
Encompass_Preferred
This damping foil consists of a dead soft aluminum foil backing coated with a pressure-sensitive viscoelastic acrylic polymer adhesive. It is engineered to control resonant vibrations and reduce structure-borne noise and fatigue in metal and composite panels and support structures. The polymer converts vibrational energy into negligible heat that dissipates, providing effective damping with as little as 20% surface coverage.
  • Features a linered construction suitable for die-cutting applications.
  • Provides excellent resistance to weathering, moisture, and UV degradation.
  • Meets F.A.R. 25.853(a) flame retardancy requirements.
  • Effective damping across temperatures from -25°F to 175°F (-32°C to 80°C), with peak performance from 32°F to 140°F (0°C to 60°C).
  • Suitable for industrial, aerospace, automotive, and appliance applications.
Catalog No. 50-275-7057 Supplier Grainger Supplier No. 15C974
This damping foil consists of a dead soft aluminum foil backing coated with a pressure-sensitive viscoelastic acrylic polymer adhesive. It is engineered to control resonant vibrations and reduce structure-borne noise and fatigue in metal and composite panels and support structures. The polymer converts vibrational energy into negligible heat that dissipates, providing effective damping with as little as 20% surface coverage.
  • Features a linered construction suitable for die-cutting applications.
  • Provides excellent resistance to weathering, moisture, and UV degradation.
  • Meets F.A.R. 25.853(a) flame retardancy requirements.
  • Effective damping across temperatures from -25°F to 175°F (-32°C to 80°C), with peak performance from 32°F to 140°F (0°C to 60°C).
  • Suitable for industrial, aerospace, automotive, and appliance applications.