This glass cloth reinforced aluminum foil tape is engineered for high-temperature, heat-reflective protective wrapping of cables, instruments, and other components in aerospace and industrial environments. Featuring a pressure-sensitive silicone adhesive, the tape provides reliable bonding to irregular surfaces and resists flame, moisture, weather, and UV degradation. It offers high tensile strength with superior puncture and tear resistance, maintaining consistent performance across extreme temperatures.
- Thermal resistance: Performs continuously from -54°C (-65°F) to 316°C (600°F).
- Durable construction: Reinforced glass cloth and aluminum foil backing provide high strength and puncture resistance.
- High conformability: Lightweight, flexible design conforms to curved or irregular substrates.
- Meets F.A.R. 25.853 (a) requirements.