Learn More
Grainger Tapecase Foil Tape, Stainless Steel, Acrylic Adhesive, 2 in x 3 yd, 1 ea
Catalog No. 502757251
Encompass_Preferred
This stainless steel foil tape is designed for corrosion-resistant sealing and repair in demanding environments. Featuring an acrylic adhesive on a nonmagnetic stainless steel backing, it is suitable for use in food processing plants, marine facilities, and nuclear environments, or for patching scratched surfaces.
  • Stainless steel foil backing construction
  • Corrosion-resistant properties
  • Single-sided adhesive format
  • Temperature performance range of -30°F to 310°F
  • 4 mil total tape thickness
  • Metal detectable
  • Includes polycoated kraft paper liner
  • Compliant with MIL and MIL-S-5059 standards
Catalog No. 50-275-7251 Supplier Grainger Supplier No. 15D570
This stainless steel foil tape is designed for corrosion-resistant sealing and repair in demanding environments. Featuring an acrylic adhesive on a nonmagnetic stainless steel backing, it is suitable for use in food processing plants, marine facilities, and nuclear environments, or for patching scratched surfaces.
  • Stainless steel foil backing construction
  • Corrosion-resistant properties
  • Single-sided adhesive format
  • Temperature performance range of -30°F to 310°F
  • 4 mil total tape thickness
  • Metal detectable
  • Includes polycoated kraft paper liner
  • Compliant with MIL and MIL-S-5059 standards