Learn More
Grainger 3M™ Aluminum Foil Tape, 3.1 mil, 2 in. Width, 5 yd. Length
Catalog No. 502749557
Encompass_Preferred
This dead soft aluminum foil tape features a pressure-sensitive acrylic adhesive, designed for thermal conductivity, heat and light reflection, and chemical resistance. Its conformable backing allows for reliable performance on curved and uneven surfaces in both indoor and outdoor environments.
  • Dead soft aluminum foil backing conforms to irregular surfaces
  • Highly engineered acrylic adhesive offers long-term durability and clean removal
  • Excellent thermal and light reflectivity for temperature-sensitive materials
  • Resistant to flame, moisture, weather, UV, and solvents
  • Suitable for electroplating maskant applications without bath contamination
  • Meets FAR 25.853(a) requirements and is UL 723 Class L listed
  • Operating temperature range: -54°C to +149°C (-65°F to +300°F)
Catalog No. 50-274-9557 Supplier Grainger Supplier No. 15D120
This dead soft aluminum foil tape features a pressure-sensitive acrylic adhesive, designed for thermal conductivity, heat and light reflection, and chemical resistance. Its conformable backing allows for reliable performance on curved and uneven surfaces in both indoor and outdoor environments.
  • Dead soft aluminum foil backing conforms to irregular surfaces
  • Highly engineered acrylic adhesive offers long-term durability and clean removal
  • Excellent thermal and light reflectivity for temperature-sensitive materials
  • Resistant to flame, moisture, weather, UV, and solvents
  • Suitable for electroplating maskant applications without bath contamination
  • Meets FAR 25.853(a) requirements and is UL 723 Class L listed
  • Operating temperature range: -54°C to +149°C (-65°F to +300°F)