Learn More
Grainger 3M High Temperature Aluminum Foil Tape, Silver, 2 in. Wide, 1 ea.
Catalog No. 502750975
Encompass_Preferred
This high-temperature aluminum foil tape features a dead soft backing and a silicone pressure-sensitive adhesive designed for demanding heat-shielding, splicing, and insulation applications. The tape is thermally conductive to help dissipate heat and act as a moisture-resistant vapor barrier. The adhesive gradually thermosets at high temperatures, becoming firm to maintain a secure hold.
  • Dead soft backing conforms to curved and uneven surfaces
  • Operates in a temperature range of -65°F to 600°F (-54°C to 316°C)
  • Flame-resistant and meets F.A.R. 25.853(a) requirements
  • UL-recognized component (file QQQW2.MH122798)
  • Thermally conductive for heat dissipation and improved thermal efficiency
  • Metal detectable
Catalog No. 50-275-0975 Supplier Grainger Supplier No. 15D125
This high-temperature aluminum foil tape features a dead soft backing and a silicone pressure-sensitive adhesive designed for demanding heat-shielding, splicing, and insulation applications. The tape is thermally conductive to help dissipate heat and act as a moisture-resistant vapor barrier. The adhesive gradually thermosets at high temperatures, becoming firm to maintain a secure hold.
  • Dead soft backing conforms to curved and uneven surfaces
  • Operates in a temperature range of -65°F to 600°F (-54°C to 316°C)
  • Flame-resistant and meets F.A.R. 25.853(a) requirements
  • UL-recognized component (file QQQW2.MH122798)
  • Thermally conductive for heat dissipation and improved thermal efficiency
  • Metal detectable