Learn More
Grainger 3M Damping Foil Tape, Aluminum Foil, 2 in. x 5 yd., 1 ea.
Catalog No. 502757669
Encompass_Preferred
This damping foil tape consists of a dead soft aluminum foil constraining layer coated with a pressure-sensitive, viscoelastic acrylic polymer adhesive. This construction is designed to control resonant vibrations in metal and composite structures, effectively converting vibrational energy into negligible heat. It helps reduce structure-borne noise and fatigue, providing effective damping even with minimal surface coverage.
  • Features a viscoelastic acrylic polymer adhesive for high-energy dissipation
  • Provides resonance control in temperatures from -25°F to +175°F (-32°C to +80°C)
  • Resists moisture, weather, UV degradation, and flame
  • Meets F.A.R. 25.853(a) flame retardancy requirements
  • Includes a polycoated kraft paper liner for easy die-cutting
  • Adhesion to steel is 65 oz./in. and tensile strength is 126 lbs./in.
Catalog No. 50-275-7669 Supplier Grainger Supplier No. 15C975
This damping foil tape consists of a dead soft aluminum foil constraining layer coated with a pressure-sensitive, viscoelastic acrylic polymer adhesive. This construction is designed to control resonant vibrations in metal and composite structures, effectively converting vibrational energy into negligible heat. It helps reduce structure-borne noise and fatigue, providing effective damping even with minimal surface coverage.
  • Features a viscoelastic acrylic polymer adhesive for high-energy dissipation
  • Provides resonance control in temperatures from -25°F to +175°F (-32°C to +80°C)
  • Resists moisture, weather, UV degradation, and flame
  • Meets F.A.R. 25.853(a) flame retardancy requirements
  • Includes a polycoated kraft paper liner for easy die-cutting
  • Adhesion to steel is 65 oz./in. and tensile strength is 126 lbs./in.