Learn More
Grainger 3M Aluminum Foil Tape, Acrylic Adhesive, 2 in. x 5 yd., 1 ea
Catalog No. 502748791
Encompass_Preferred
This aluminum foil tape features a dead soft aluminum foil backing coated with a highly engineered acrylic adhesive. It is designed to withstand flame, weathering, moisture, and UV, making it ideal for industrial applications including heat shielding, light enhancement, and chemical masking. The tape is thermally conductive to help dissipate heat and improve efficiency, while maintaining a strong bond in environments from -65 °F to +300 °F.
  • Dead soft aluminum foil backing conforms to curved or uneven surfaces
  • Highly engineered acrylic adhesive provides long-term durability and clean removal
  • Thermally conductive properties reduce hot spots and protect substrates
  • Resistant to flame, moisture, and weathering for indoor or outdoor use
  • Recognized by UL 746C and classified per UL 723 Class L
  • Meets F.A.R 25.853(a) and L-T-80C specifications
Catalog No. 50-274-8791 Supplier Grainger Supplier No. 15D115
This aluminum foil tape features a dead soft aluminum foil backing coated with a highly engineered acrylic adhesive. It is designed to withstand flame, weathering, moisture, and UV, making it ideal for industrial applications including heat shielding, light enhancement, and chemical masking. The tape is thermally conductive to help dissipate heat and improve efficiency, while maintaining a strong bond in environments from -65 °F to +300 °F.
  • Dead soft aluminum foil backing conforms to curved or uneven surfaces
  • Highly engineered acrylic adhesive provides long-term durability and clean removal
  • Thermally conductive properties reduce hot spots and protect substrates
  • Resistant to flame, moisture, and weathering for indoor or outdoor use
  • Recognized by UL 746C and classified per UL 723 Class L
  • Meets F.A.R 25.853(a) and L-T-80C specifications