Learn More
Grainger 3M Aluminum Foil Tape, 3 in. Wide, 5 yd. Long, Silver, 1 ea.
Catalog No. 502750972
Encompass_Preferred
This aluminum foil tape is designed for various sealing, holding, splicing, and masking applications across multiple industries, including aerospace and manufacturing. It features a dead-soft aluminum foil backing with a transparent acrylic adhesive, providing excellent reflective properties for heat and light. This tape performs well in harsh indoor and outdoor environments and offers a low moisture vapor transmission rate for effective sealing.
  • Provides excellent heat and light reflection
  • Offers good long-term aging performance indoors and outdoors
  • Features low moisture vapor transmission for reliable sealing and patching
  • Flame resistant and UL 723 class L rated for low flammability
  • Meets FAR 25.853(a) requirements for use in commercial aircraft
  • Compatible with a wide temperature range of -65°F to 300°F (-53°C to 148°C)
  • Metal detectable
Catalog No. 50-275-0972 Supplier Grainger Supplier No. 15D121
This aluminum foil tape is designed for various sealing, holding, splicing, and masking applications across multiple industries, including aerospace and manufacturing. It features a dead-soft aluminum foil backing with a transparent acrylic adhesive, providing excellent reflective properties for heat and light. This tape performs well in harsh indoor and outdoor environments and offers a low moisture vapor transmission rate for effective sealing.
  • Provides excellent heat and light reflection
  • Offers good long-term aging performance indoors and outdoors
  • Features low moisture vapor transmission for reliable sealing and patching
  • Flame resistant and UL 723 class L rated for low flammability
  • Meets FAR 25.853(a) requirements for use in commercial aircraft
  • Compatible with a wide temperature range of -65°F to 300°F (-53°C to 148°C)
  • Metal detectable