Learn More
Grainger Tapecase Foil Tape, Aluminum, 3 in x 5 yd, 4 mil, Silicone, 1 ea
Catalog No. 502756884
Encompass_Preferred
This aluminum foil tape is reinforced with glass cloth to provide a strong, flexible, and abrasion-resistant solution for demanding applications. Designed for use in both indoor and outdoor environments, it conforms effectively to curves and uneven surfaces, making it suitable for high-temperature equipment protection and insulation in industries such as aerospace and automotive.
  • Features a silicone adhesive for high-temperature performance
  • Suitable for temperatures ranging from -60°F to +600°F
  • Reinforced with glass cloth for added durability
  • Metal detectable construction
  • Suitable for use in automotive, aviation, and general maintenance applications
Catalog No. 50-275-6884 Supplier Grainger Supplier No. 15C645
This aluminum foil tape is reinforced with glass cloth to provide a strong, flexible, and abrasion-resistant solution for demanding applications. Designed for use in both indoor and outdoor environments, it conforms effectively to curves and uneven surfaces, making it suitable for high-temperature equipment protection and insulation in industries such as aerospace and automotive.
  • Features a silicone adhesive for high-temperature performance
  • Suitable for temperatures ranging from -60°F to +600°F
  • Reinforced with glass cloth for added durability
  • Metal detectable construction
  • Suitable for use in automotive, aviation, and general maintenance applications