Learn More
Grainger 3M Aluminum Foil Glass Cloth Tape 363, Silver, 1 in.
Catalog No. 502758027
Encompass_Preferred
This high-temperature aluminum foil and glass cloth tape is engineered for demanding industrial and aerospace applications requiring robust heat shielding and structural integrity. Featuring a glass cloth-reinforced foil backing and a pressure-sensitive silicone adhesive, the tape provides superior tensile strength and conformability. It effectively resists flame, moisture, weather, and UV degradation, performing consistently in temperatures ranging from -65°F to 600°F (-54°C to 316°C).
  • High tensile strength for puncture and tear resistance
  • Excellent thermal conductivity and heat reflectivity
  • Conforms effectively to irregular or curved surfaces
  • Adhesive removes cleanly from most substrates
  • Meets F.A.R. 25.853(a) flammability standards
Catalog No. 50-275-8027 Supplier Grainger Supplier No. 15C129
This high-temperature aluminum foil and glass cloth tape is engineered for demanding industrial and aerospace applications requiring robust heat shielding and structural integrity. Featuring a glass cloth-reinforced foil backing and a pressure-sensitive silicone adhesive, the tape provides superior tensile strength and conformability. It effectively resists flame, moisture, weather, and UV degradation, performing consistently in temperatures ranging from -65°F to 600°F (-54°C to 316°C).
  • High tensile strength for puncture and tear resistance
  • Excellent thermal conductivity and heat reflectivity
  • Conforms effectively to irregular or curved surfaces
  • Adhesive removes cleanly from most substrates
  • Meets F.A.R. 25.853(a) flammability standards