Learn More
Grainger 3M Shielding Foil Tape, Deadsoft Aluminum, 3 in. x 5 yd. 1 ea
Catalog No. 502748793
Encompass_Preferred
This shielding foil tape is designed for a wide range of applications in assembly, manufacturing, automotive, aviation, construction, and electronics. Constructed with a deadsoft aluminum backing and a single-sided acrylic adhesive, this durable tape provides reliable performance in both indoor and outdoor environments.
  • Deadsoft aluminum backing offers excellent conformability
  • Acrylic adhesive provides long-term bond strength
  • Heat-resistant for performance up to 300°F
  • Chemical and UV-resistant properties for harsh conditions
  • Compatible with diverse industrial applications including maintenance and repair
  • Continuous roll format measuring 76 mm (3 in.) wide by 4.5 m (5 yd.) long
Catalog No. 50-274-8793 Supplier Grainger Supplier No. 15D116
This shielding foil tape is designed for a wide range of applications in assembly, manufacturing, automotive, aviation, construction, and electronics. Constructed with a deadsoft aluminum backing and a single-sided acrylic adhesive, this durable tape provides reliable performance in both indoor and outdoor environments.
  • Deadsoft aluminum backing offers excellent conformability
  • Acrylic adhesive provides long-term bond strength
  • Heat-resistant for performance up to 300°F
  • Chemical and UV-resistant properties for harsh conditions
  • Compatible with diverse industrial applications including maintenance and repair
  • Continuous roll format measuring 76 mm (3 in.) wide by 4.5 m (5 yd.) long