Learn More
Grainger 3M Aluminum Foil Tape 425, Dead Soft Aluminum, 25 mm, 55 m
Catalog No. 502749530
Encompass_Preferred
This conductive aluminum foil tape features a dead soft foil backing paired with a pressure-sensitive acrylic adhesive. Designed for versatility, it excels in heat and light reflection, chemical masking, and thermal dissipation across a temperature range of -54°C to +149°C. The conformable backing allows for effective application on irregular surfaces, ensuring reliability in both indoor and outdoor environments.
  • Dead soft aluminum backing conforms to curved and uneven surfaces
  • Highly engineered acrylic adhesive provides long-term durability and removes cleanly after masking
  • Thermally conductive design aids in heat dissipation and improves efficiency
  • Excellent reflection of heat and light protects sensitive components
  • Flame, chemical, weathering, moisture, and UV resistant
  • Recognized by UL to UL 746C and classified to UL 723, Class L for low flammability
Catalog No. 50-274-9530 Supplier Grainger Supplier No. 15D117
This conductive aluminum foil tape features a dead soft foil backing paired with a pressure-sensitive acrylic adhesive. Designed for versatility, it excels in heat and light reflection, chemical masking, and thermal dissipation across a temperature range of -54°C to +149°C. The conformable backing allows for effective application on irregular surfaces, ensuring reliability in both indoor and outdoor environments.
  • Dead soft aluminum backing conforms to curved and uneven surfaces
  • Highly engineered acrylic adhesive provides long-term durability and removes cleanly after masking
  • Thermally conductive design aids in heat dissipation and improves efficiency
  • Excellent reflection of heat and light protects sensitive components
  • Flame, chemical, weathering, moisture, and UV resistant
  • Recognized by UL to UL 746C and classified to UL 723, Class L for low flammability