Learn More
Grainger 3M Heavy Duty Aluminum Foil Tape, 4 in, 7.2 mil, Silver
Catalog No. 502750402
Encompass_Preferred
This heavy duty aluminum foil tape features a dead soft aluminum foil backing with a transparent acrylic adhesive, designed for demanding industrial applications. The tape provides excellent thermal conductivity and serves as an effective heat and light reflective surface. It is engineered to withstand exposure to flame, weathering, moisture, and UV, providing a durable service life for both indoor and outdoor environments.
  • Meets UL 723 Class L low flammability rating and FAR 25.853(a) requirements
  • Performs in a temperature range from -65°F to 300°F
  • Features a 7.2 mil total thickness with 5.0 mil foil backing
  • Allows expansion and contraction with temperature changes due to high coefficient of expansion
  • Resistant to moisture and UV exposure
  • Provides an excellent reflective surface for heat and light
Catalog No. 50-275-0402 Supplier Grainger Supplier No. 15D129
This heavy duty aluminum foil tape features a dead soft aluminum foil backing with a transparent acrylic adhesive, designed for demanding industrial applications. The tape provides excellent thermal conductivity and serves as an effective heat and light reflective surface. It is engineered to withstand exposure to flame, weathering, moisture, and UV, providing a durable service life for both indoor and outdoor environments.
  • Meets UL 723 Class L low flammability rating and FAR 25.853(a) requirements
  • Performs in a temperature range from -65°F to 300°F
  • Features a 7.2 mil total thickness with 5.0 mil foil backing
  • Allows expansion and contraction with temperature changes due to high coefficient of expansion
  • Resistant to moisture and UV exposure
  • Provides an excellent reflective surface for heat and light