Learn More
Grainger Foil Tape 6 in x 5 yd Aluminum
Catalog No. 502750973
Encompass_Preferred
The 3M Foil Tape 15D123 is a 2" wide, 60 yard long aluminum foil tape with acrylic adhesive. It has a temperature resistance range of -20°F to 250°F and is designed for HVAC sealing and joining applications. This UL listed tape provides an air and moisture barrier and can be used for ductwork fabrication and repair. It bonds well to sheet metal, fiberglass and other common ductwork materials. The acrylic adhesive adheres quickly and provides a long lasting bond.
Catalog No. 50-275-0973 Supplier Grainger Supplier No. 15D123
The 3M Foil Tape 15D123 is a 2" wide, 60 yard long aluminum foil tape with acrylic adhesive. It has a temperature resistance range of -20°F to 250°F and is designed for HVAC sealing and joining applications. This UL listed tape provides an air and moisture barrier and can be used for ductwork fabrication and repair. It bonds well to sheet metal, fiberglass and other common ductwork materials. The acrylic adhesive adheres quickly and provides a long lasting bond.