Learn More
Grainger Foil Tape 6 in x 5 yd Aluminum
Catalog No. 502752831
Encompass_Preferred
The 3M Foil Tape 15D130 is a 2 inch wide, 60 yard long aluminum foil tape with acrylic adhesive. It has a temperature resistance from -20F to 250F and is designed for HVAC sealing and joining applications. This UL listed tape provides a strong bond and moisture seal while conforming to irregular surfaces. It can be used for sealing joints and seams on foil-faced insulation materials and duct work. The acrylic adhesive adheres well to sheet metal, glass, wood, plastic, and more. This durable tape resists UV light and withstands repeated freezing and thawing.
Catalog No. 50-275-2831 Supplier Grainger Supplier No. 15D130
The 3M Foil Tape 15D130 is a 2 inch wide, 60 yard long aluminum foil tape with acrylic adhesive. It has a temperature resistance from -20F to 250F and is designed for HVAC sealing and joining applications. This UL listed tape provides a strong bond and moisture seal while conforming to irregular surfaces. It can be used for sealing joints and seams on foil-faced insulation materials and duct work. The acrylic adhesive adheres well to sheet metal, glass, wood, plastic, and more. This durable tape resists UV light and withstands repeated freezing and thawing.