Learn More
Research Products International Corp FoilSeal RollMate Film DIS w/ Pre-Scored Sheets, 500/rl
SDP
Catalog No. 502230147
Encompass
FoilSeal RollMate Film is an economical, efficient way to rapidly seal microplates. Pre-scored foil sheets offer a faster method of applying and sealing plates. Easy to use roll format is convenient for PCR, long-term/cold storage, light sensitive assays and robotics. Self-standing dispenser features non-slip supports that hold pre-scored foil roll securely in place for quick and easy application.
Chemically resistant film is 2mil thick, pierceable and conforms to wells for visual verification and positive seal. Specially formulated adhesive provides excellent seal, minimizes evaporation, reduce cros-contamination, and removes easily and cleanly from plate surface. Supplied 500 per roll. FoilSeal is a Trademark of Excel Scientific, Inc.
Easy to Use Roll
DMSO Resistant
Withstand -80°C to +120°C;
RNase, DNase free
Catalog No. 50-223-0147 Supplier Research Products International Corp Supplier No. 202510
FoilSeal RollMate Film is an economical, efficient way to rapidly seal microplates. Pre-scored foil sheets offer a faster method of applying and sealing plates. Easy to use roll format is convenient for PCR, long-term/cold storage, light sensitive assays and robotics. Self-standing dispenser features non-slip supports that hold pre-scored foil roll securely in place for quick and easy application.
Chemically resistant film is 2mil thick, pierceable and conforms to wells for visual verification and positive seal. Specially formulated adhesive provides excellent seal, minimizes evaporation, reduce cros-contamination, and removes easily and cleanly from plate surface. Supplied 500 per roll. FoilSeal is a Trademark of Excel Scientific, Inc.
Easy to Use Roll
DMSO Resistant
Withstand -80°C to +120°C;
RNase, DNase free