Learn More
New England Biolabs, Inc. FokI – 5000 units
SDP
Catalog No. 50811942
Encompass
A restriction endonuclease that recognizes the sequence GGATGNNNNNNNNN^NNNN_.
Catalog No. 50-811-942 Supplier New England Biolabs, Inc. Supplier No. R0109L
A restriction endonuclease that recognizes the sequence GGATGNNNNNNNNN^NNNN_.