Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Folate-PEG3-C2-acid | 97.6% | 644.63 g/mol | C28H36N8O10 | 10 MG
SDP

Catalog No. 5000298590
Encompass_Preferred

Folate-PEG3-C2-acid is the acid fragment of a folate-PEG3-based NHS ester and serves as a PEG-type PROTAC linker building block used in the synthesis of PROTAC molecules and folate-conjugated ligands for receptor-targeting studies.

  • Purity: 97.6%.
  • Molecular weight: 644.63 g/mol.
  • Solubility: soluble in DMSO at 33.33 mg/mL (51.70 mM); ultrasonication and heating may aid dissolution.
  • Storage: store at 4°C protected from light; in solvent keep at -80°C (6 months) or -20°C (1 month), protected from light.
  • Physical state: light yellow to yellow solid.

Catalog No. 50-002-98590 Supplier Medchemexpress LLC Supplier No. HY4737310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Folate-PEG3-C2-acid is the acid fragment of a folate-PEG3-based NHS ester and serves as a PEG-type PROTAC linker building block used in the synthesis of PROTAC molecules and folate-conjugated ligands for receptor-targeting studies.

  • Purity: 97.6%.
  • Molecular weight: 644.63 g/mol.
  • Solubility: soluble in DMSO at 33.33 mg/mL (51.70 mM); ultrasonication and heating may aid dissolution.
  • Storage: store at 4°C protected from light; in solvent keep at -80°C (6 months) or -20°C (1 month), protected from light.
  • Physical state: light yellow to yellow solid.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.