Learn More
Medchemexpress LLC Folate-PEG3-C2-acid | 97.6% | C28H36N8O10 | 1 MG
SDP
Catalog No. 5000357757
Encompass_Preferred
Folate-PEG3-C2-acid is a specialized acid fragment derived from Folate-PEG3-NHS ester, designed as a PEG-type PROTAC linker. This compound is crucial for the synthesis of PROTAC (PROteolysis TArgeting Chimera) molecules, which are heterobifunctional molecules that induce the degradation of target proteins. Its molecular structure facilitates the creation of complex PROTACs, enabling advanced research in targeted protein degradation.
  • PEG-type PROTAC linker
  • Acid fragment of Folate-PEG3-NHS ester
  • Essential for synthesizing PROTAC molecules
  • Purity of 97.56% ensures reliable experimental results
Catalog No. 50-003-57757 Supplier Medchemexpress LLC Supplier No. HY473731MG
Folate-PEG3-C2-acid is a specialized acid fragment derived from Folate-PEG3-NHS ester, designed as a PEG-type PROTAC linker. This compound is crucial for the synthesis of PROTAC (PROteolysis TArgeting Chimera) molecules, which are heterobifunctional molecules that induce the degradation of target proteins. Its molecular structure facilitates the creation of complex PROTACs, enabling advanced research in targeted protein degradation.
  • PEG-type PROTAC linker
  • Acid fragment of Folate-PEG3-NHS ester
  • Essential for synthesizing PROTAC molecules
  • Purity of 97.56% ensures reliable experimental results