Learn More
Biocare Medical, LLC Folate Receptor alpha IHC Assay Kit Antibody - Predilute, 32 Tests
SDP
Catalog No. NC0283348
Encompass_Preferred
Mouse anti-human folate receptor alpha antibody monoclonal [26B3.F2] specifically recognizes the alpha isoform of folate receptor. Folate receptor alpha is primarily expressed in the apical surface of some polarized epithelial cells of normal tissues (such as kidney, lung, breast, and salivary glands) and on many cancer cells of epithelial origin; namely, ovarian cancer, thyroid cancer, non-small cell lung adenocarcinoma (NSCLC) and endometrial cancer.In the most widely studied tumor, epithelial ovarian cancer, expression of FRalpha increases with tumor stage [12], and is associated with decreased survival [11]. However, in NSCLC, FRalpha has been shown to be specific for adenocarcinomas relative to squamous cell carcinoma and other histologic subtypes, and increased expression has been correlated to increased survival [4,9].
Catalog No. NC0283348 Supplier Biocare Medical, LLC Supplier No. BRI4006KAA
Mouse anti-human folate receptor alpha antibody monoclonal [26B3.F2] specifically recognizes the alpha isoform of folate receptor. Folate receptor alpha is primarily expressed in the apical surface of some polarized epithelial cells of normal tissues (such as kidney, lung, breast, and salivary glands) and on many cancer cells of epithelial origin; namely, ovarian cancer, thyroid cancer, non-small cell lung adenocarcinoma (NSCLC) and endometrial cancer.In the most widely studied tumor, epithelial ovarian cancer, expression of FRalpha increases with tumor stage [12], and is associated with decreased survival [11]. However, in NSCLC, FRalpha has been shown to be specific for adenocarcinomas relative to squamous cell carcinoma and other histologic subtypes, and increased expression has been correlated to increased survival [4,9].