Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Foley Catheter Holster
SDP

Catalog No. 22000689
Change view
Click to view available options
Length (English):
16 in.
22 in.
Includes:
Magnet
Tape
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Includes
22-000-689 22 in. Tape
22-000-686 16 in. Magnet
22-000-687 16 in. Tape
22-000-688 22 in. Magnet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 22-000-689 Supplier Poltex Supplier No. FLYHLST22T
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Poltex Foley Catheter Holster is used to access catheters as well as aides in cleaning.

  • Foley Catheter holster.
  • Mounts to side of procedure cart or other convenient space.
  • Removable caddy helps to access catheters as well as aides in cleaning.
  • Bottom hole is closed when in use, but opens for easy cleaning.
  • Top lid helps to prevent dust accumulation.
  • Cleanable PETG plastic construction.
  • Choose from 16.3 in. internal height and 22.3 in. internal height.
  • Choose from powerful VHB Tape mount, or powerful magnet mount.

Specifications

Includes Tape
Length (English) 22 in.
Product Type Foley Catheter Holder
Disposable Reusable
For Use With (Equipment) Indwelling Catheter
Graduated No
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.