Learn More
Enzo Life Sciences Folic acid (10g). CAS: 59-30-3
SDP
Catalog No. 502009285
Encompass_Preferred
Alternative name: Pteroylglutamic acid. Purity: ≥97% (HPLC). Solubility: Soluble in diluted acids or alkaline solutions; slightly soluble in water or methanol; almost insoluble in organic solvents. Long Term Storage: +4°C.
Catalog No. 50-200-9285 Supplier Enzo Life Sciences Supplier No. ALX460006G010
Alternative name: Pteroylglutamic acid. Purity: ≥97% (HPLC). Solubility: Soluble in diluted acids or alkaline solutions; slightly soluble in water or methanol; almost insoluble in organic solvents. Long Term Storage: +4°C.