Learn More
Medchemexpress LLC Folic acid-13C5 | 1207282-75-4 | 95.0% | 446.36 | 500 UG
SDP
Catalog No. 5000394698
Encompass_Preferred
Folic acid-13C5 is the 13C-labeled Folic acid. Folic acid (Vitamin M; Vitamin B9) is a B vitamin; it is necessary for the production and maintenance of new cells, for DNA synthesis and RNA synthesis. This product is for research use only and not sold to patients.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-94698 Supplier Medchemexpress LLC Supplier No. HY16637S3500UG
Folic acid-13C5 is the 13C-labeled Folic acid. Folic acid (Vitamin M; Vitamin B9) is a B vitamin; it is necessary for the production and maintenance of new cells, for DNA synthesis and RNA synthesis. This product is for research use only and not sold to patients.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.