Learn More
U.S. Pharmacopeia Folic Acid, 59-30-3, MFCD00079305, 500mg
Supplier:  U.S. Pharmacopeia 1286005
Encompass
Molecular formula C19H19N7O6, Molecular weight 441.45 g/mol, Softens, darkens, and chars above 250 ° C, Synonyms: Pteroylglutamic acid, Vitamin B9
Catalog No. NC0088411