Learn More
Apexbio Technology LLC Folic acid 59-30-3 10mM (in 1mL DMSO)
SDP
Catalog No. 5000093028
Encompass_Preferred
Folic acid (CAS 59-30-3) is a water-soluble B9 vitamin structurally derived from Spinacia oleracea It functions as a cofactor in one-carbon transfer reactions facilitating nucleotide synthesis and methylation processes essential for DNA replication and cellular division Due to its involvement in folate metabolism folic acid is frequently utilized in studies of cell proliferation gene expression regulation and metabolic disorders For optimal stability folic acid should be stored in a sealed container under cool dry conditions
Catalog No. 50-000-93028 Supplier Apexbio Technology LLC Supplier No. N20755.1
Folic acid (CAS 59-30-3) is a water-soluble B9 vitamin structurally derived from Spinacia oleracea It functions as a cofactor in one-carbon transfer reactions facilitating nucleotide synthesis and methylation processes essential for DNA replication and cellular division Due to its involvement in folate metabolism folic acid is frequently utilized in studies of cell proliferation gene expression regulation and metabolic disorders For optimal stability folic acid should be stored in a sealed container under cool dry conditions