Learn More
Medchemexpress LLC Folic acid | 59-30-3 | 99.0% | 441.40 | 1 ML
SDP
Catalog No. 5000392391
Encompass_Preferred
Folic acid is an orally active essential nutrient from the B complex group of vitamins. It has shown antidepressant-like effects and can reduce the risk of neonatal neural tube defects. It can also be used in research for megaloblastic and macrocytic anemias due to its deficiency.
  • An orally active essential nutrient from the B complex group of vitamins.
  • Shows antidepressant-like effects.
  • Reduces the risk of neonatal neural tube defects.
  • Can be used to research megaloblastic and macrocytic anemias due to its deficiency.
Catalog No. 50-003-92391 Supplier Medchemexpress LLC Supplier No. HY1663710MM/1ML
Folic acid is an orally active essential nutrient from the B complex group of vitamins. It has shown antidepressant-like effects and can reduce the risk of neonatal neural tube defects. It can also be used in research for megaloblastic and macrocytic anemias due to its deficiency.
  • An orally active essential nutrient from the B complex group of vitamins.
  • Shows antidepressant-like effects.
  • Reduces the risk of neonatal neural tube defects.
  • Can be used to research megaloblastic and macrocytic anemias due to its deficiency.