Learn More
MOLECULAR DEPOT LLC Folic Acid Diluent - 50 mL
Catalog No. 5000013462
Encompass_Preferred
Folic Acid Buffered Diluent is a liquid-stable buffered solution containing undetectable levels of folate (0 ng/mL) This product has been used in the research and development of folic acid high and low working solutions This product is for in vitro research use only
Catalog No. 50-000-13462 Supplier MOLECULAR DEPOT LLC Supplier No. B2010581
Folic Acid Buffered Diluent is a liquid-stable buffered solution containing undetectable levels of folate (0 ng/mL) This product has been used in the research and development of folic acid high and low working solutions This product is for in vitro research use only