Learn More
Medchemexpress LLC Folic acid (disodium) | 29704-76-5 | 98.6% | 485.36 | 25 MG
SDP
Catalog No. 5000393016
Encompass_Preferred
Folic acid disodium (Vitamin B9 disodium; Vitamin M disodium) is an orally active disodium salt form of Folic acid with an intrinsic dissolution rate (IDR) of 4.96·10^5 g/s. It serves as a cofactor in single-carbon transfer reactions and exhibits protective effects against neural tube defects, ischemic events, and cancer. Folate acid disodium overload leads to impaired brain development in embryogenesis and promotes growth of precancerous altered cells. Folic acid deficiency leads to megaloblastic anemia.
  • Orally active disodium salt form of folic acid.
  • Serves as cofactor in single-carbon transfer reactions.
  • Exhibits protective effects against neural tube defects, ischemic events, and cancer.
  • Deficiency leads to megaloblastic anemia.
Catalog No. 50-003-93016 Supplier Medchemexpress LLC Supplier No. 25MGHY16637D
Folic acid disodium (Vitamin B9 disodium; Vitamin M disodium) is an orally active disodium salt form of Folic acid with an intrinsic dissolution rate (IDR) of 4.96·10^5 g/s. It serves as a cofactor in single-carbon transfer reactions and exhibits protective effects against neural tube defects, ischemic events, and cancer. Folate acid disodium overload leads to impaired brain development in embryogenesis and promotes growth of precancerous altered cells. Folic acid deficiency leads to megaloblastic anemia.
  • Orally active disodium salt form of folic acid.
  • Serves as cofactor in single-carbon transfer reactions.
  • Exhibits protective effects against neural tube defects, ischemic events, and cancer.
  • Deficiency leads to megaloblastic anemia.