Learn More
Medchemexpress LLC Folic acid disodium | 29704-76-5 | 98.6% | 485.36 | 50 MG
SDP
Catalog No. 5000392395
Encompass_Preferred
Folic acid disodium, also known as Vitamin B9 disodium or Vitamin M disodium, is an orally active disodium salt form of Folic acid. It functions as a cofactor in single-carbon transfer reactions and provides protective effects against neural tube defects, ischemic events, and certain cancers. Conversely, an overload of folic acid disodium can impair brain development during embryogenesis and promote the growth of precancerous altered cells, and a deficiency can lead to megaloblastic anemia.
  • Orally active disodium salt form of folic acid.
  • Acts as a cofactor in single-carbon transfer reactions.
  • Offers protective effects against neural tube defects, ischemic events, and certain cancers.
  • High purity of 98.56%.
  • Molecular weight of 485.36.
Catalog No. 50-003-92395 Supplier Medchemexpress LLC Supplier No. HY16637D50MG
Folic acid disodium, also known as Vitamin B9 disodium or Vitamin M disodium, is an orally active disodium salt form of Folic acid. It functions as a cofactor in single-carbon transfer reactions and provides protective effects against neural tube defects, ischemic events, and certain cancers. Conversely, an overload of folic acid disodium can impair brain development during embryogenesis and promote the growth of precancerous altered cells, and a deficiency can lead to megaloblastic anemia.
  • Orally active disodium salt form of folic acid.
  • Acts as a cofactor in single-carbon transfer reactions.
  • Offers protective effects against neural tube defects, ischemic events, and certain cancers.
  • High purity of 98.56%.
  • Molecular weight of 485.36.