Learn More
Biomatik Corporation Folic Acid (FA) CLIA Kit
Supplier:  Biomatik Corporation EKU11560
Encompass_Preferred
Folic Acid (FA) CLIA Kit for detecting FA in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Universal. Detection Range: 39.1-10,000pg/ml. Sensitivity: 16.4pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Target Synonyms: VB9; Vitamin B9; Folacin; Folate; V; Itamin M; Vitamin M; Vitamin Bc; Pteroyl-L-Glutamic Acid; Pteroyl-L-Glutamate. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8123