Learn More
Abcam Folic acid (Folate), Synthetic vitamin B9, 25G
Catalog No. 502706098
Encompass_Preferred
MW 441.4 Da, Purity >96%. Synthetic vitamin B9. Tetrahydrofolate precursor. Promotes DNA synthesis and cell division. Shows anti-inflammatory and chemopreventive effects *in vivo.* Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6098 Supplier Abcam Supplier No. AB14362125G
MW 441.4 Da, Purity >96%. Synthetic vitamin B9. Tetrahydrofolate precursor. Promotes DNA synthesis and cell division. Shows anti-inflammatory and chemopreventive effects *in vivo.* Orally active.
The product is subject to the following: Abcam Restricted Use Statement