Learn More
MOLECULAR DEPOT LLC Folic Acid Magnetic - 1 mL
Catalog No. 5000013368
Encompass_Preferred
Folic Acid Magnetic Beads consists of a suspension of magnetic particles (3 um) conjugated to Folic Acid (Folate) Highly specific and highly stable these particles can be used for the easy one-step capture of various antigens proteins and antibodies specific to Folic Acid (Folate) These particles can be recovered easily using a magnet allowing for various applications such as purification depletion assay development and more For in vitro research use only
Catalog No. 50-000-13368 Supplier MOLECULAR DEPOT LLC Supplier No. B2010397
Folic Acid Magnetic Beads consists of a suspension of magnetic particles (3 um) conjugated to Folic Acid (Folate) Highly specific and highly stable these particles can be used for the easy one-step capture of various antigens proteins and antibodies specific to Folic Acid (Folate) These particles can be recovered easily using a magnet allowing for various applications such as purification depletion assay development and more For in vitro research use only