Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Folic acid-PEG5-Mal | 00-00-0 | 99.5% | C35H45N9O12 | 10MG
SDP

Catalog No. 5000261037
Encompass_Preferred

FA-PEG5-Mal is a polyethylene glycol (PEG)-based linker developed for constructing proteolysis-targeting chimeras (PROTACs). It combines a folic acid-derived ligand with a PEG5 spacer and a maleimide functional group to enable selective conjugation to thiol-containing partners. Supplied as a solid for laboratory research use, it offers high purity and convenient solubility in DMSO for bioconjugation and linker assembly workflows.

  • Provides a folic acid ligand for target engagement
  • Contains a PEG5 spacer to give flexible linker length
  • Includes a maleimide functional handle for thiol coupling
  • Supplied as a solid suitable for laboratory research use
  • High purity for reliable synthetic results
  • Soluble in DMSO (100 mg/mL) with ultrasonic assistance recommended

Catalog No. 50-002-61037 Supplier Medchemexpress LLC Supplier No. HY13207010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FA-PEG5-Mal is a polyethylene glycol (PEG)-based linker developed for constructing proteolysis-targeting chimeras (PROTACs). It combines a folic acid-derived ligand with a PEG5 spacer and a maleimide functional group to enable selective conjugation to thiol-containing partners. Supplied as a solid for laboratory research use, it offers high purity and convenient solubility in DMSO for bioconjugation and linker assembly workflows.

  • Provides a folic acid ligand for target engagement
  • Contains a PEG5 spacer to give flexible linker length
  • Includes a maleimide functional handle for thiol coupling
  • Supplied as a solid suitable for laboratory research use
  • High purity for reliable synthetic results
  • Soluble in DMSO (100 mg/mL) with ultrasonic assistance recommended

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.