Learn More
Medchemexpress LLC Folic acid-PEG5-maleimide | 00-00-0 | 99.5% | 783.78 g/mol | C35H45N9O12 | 5 MG
SDP
Catalog No. 5000263087
Encompass_Preferred
FA-PEG5-Mal (folic acid-PEG5-maleimide) is a polyethylene glycol-based bifunctional linker that combines a folic acid recognition unit with a terminal maleimide for thiol coupling. It is intended for use in the synthesis of PROTACs and folate-targeted conjugates, providing a water-soluble spacer to connect ligands for targeted degradation or delivery.
  • Provides a folic acid recognition unit for folate receptor targeting.
  • Features a terminal maleimide for selective thiol conjugation.
  • Uses a PEG5 spacer to impart water solubility and flexibility.
  • High purity suitable for research-grade chemical synthesis.
  • Available in small milligram packages for conjugation workflows.
  • Stable when stored sealed and protected from moisture and light; in solvent: -80°C (6 months), -20°C (1 month).
Catalog No. 50-002-63087 Supplier Medchemexpress LLC Supplier No. HY1320705MG
FA-PEG5-Mal (folic acid-PEG5-maleimide) is a polyethylene glycol-based bifunctional linker that combines a folic acid recognition unit with a terminal maleimide for thiol coupling. It is intended for use in the synthesis of PROTACs and folate-targeted conjugates, providing a water-soluble spacer to connect ligands for targeted degradation or delivery.
  • Provides a folic acid recognition unit for folate receptor targeting.
  • Features a terminal maleimide for selective thiol conjugation.
  • Uses a PEG5 spacer to impart water solubility and flexibility.
  • High purity suitable for research-grade chemical synthesis.
  • Available in small milligram packages for conjugation workflows.
  • Stable when stored sealed and protected from moisture and light; in solvent: -80°C (6 months), -20°C (1 month).