Learn More
Alkali Scientific Folic Acid [Pteroylglutamic acid] [Vitamin M], 5 G
SDP
Catalog No. 5000661364
Encompass_Preferred
Folic Acid (Pteroylglutamic acid), also known as Vitamin B9, is a compound used in cell culture applications, supplied in a 5g quantity. Folic acid is essential for DNA synthesis, repair, and cellular division, making it a vital component in maintaining cell growth and function. In cell culture, folic acid is typically used to support the growth and proliferation of eukaryotic cells, particularly in mammalian cell culture systems. It is also utilized in research related to gene expression, cellular metabolism, and the study of folate metabolism in various biological systems. The 5g packaging is ideal for laboratories requiring consistent and stable folic acid supplementation in their cell culture medium.
Catalog No. 50-006-61364 Supplier Alkali Scientific Supplier No. OLI004CP
Folic Acid (Pteroylglutamic acid), also known as Vitamin B9, is a compound used in cell culture applications, supplied in a 5g quantity. Folic acid is essential for DNA synthesis, repair, and cellular division, making it a vital component in maintaining cell growth and function. In cell culture, folic acid is typically used to support the growth and proliferation of eukaryotic cells, particularly in mammalian cell culture systems. It is also utilized in research related to gene expression, cellular metabolism, and the study of folate metabolism in various biological systems. The 5g packaging is ideal for laboratories requiring consistent and stable folic acid supplementation in their cell culture medium.