Learn More
MOLECULAR DEPOT LLC Folic acid Release - 50 mL
Catalog No. 5000013402
Encompass_Preferred
Folic Acid Release Buffer is an aqueous buffer designed to the fast and efficient release of folic acid (folate) from its serum/plasma transporters Unlike other release agents this buffer is organic solvent-free It is supplied as two liquid solutions to combine before use For in vitro research use only
Catalog No. 50-000-13402 Supplier MOLECULAR DEPOT LLC Supplier No. B2010579
Folic Acid Release Buffer is an aqueous buffer designed to the fast and efficient release of folic acid (folate) from its serum/plasma transporters Unlike other release agents this buffer is organic solvent-free It is supplied as two liquid solutions to combine before use For in vitro research use only