Learn More
Medchemexpress LLC Folic acid (standard) | 59-30-3 | 98.4% | 441.40 | 100 MG
SDP
Catalog No. 5000392190
Encompass_Preferred
Folic acid (Standard) is the analytical standard of Folic acid, intended for research and analytical applications. Folic acid (Vitamin B9) is an orally active essential nutrient from the B complex group of vitamins. It shows antidepressant-like effects and its sodium form reduces the risk of neonatal neural tube defects. It can be used in the research of megaloblastic and macrocytic anemias due to folic deficiency.
  • Analytical standard for assays
  • Used in qualitative, quantitative, and methodological research experiments
  • Shows antidepressant-like effect
  • Reduces the risk of neonatal neural tube defects (sodium form)
  • Used for research of megaloblastic and macrocytic anemias due to folic deficiency
Catalog No. 50-003-92190 Supplier Medchemexpress LLC Supplier No. HY16637R100MG
Folic acid (Standard) is the analytical standard of Folic acid, intended for research and analytical applications. Folic acid (Vitamin B9) is an orally active essential nutrient from the B complex group of vitamins. It shows antidepressant-like effects and its sodium form reduces the risk of neonatal neural tube defects. It can be used in the research of megaloblastic and macrocytic anemias due to folic deficiency.
  • Analytical standard for assays
  • Used in qualitative, quantitative, and methodological research experiments
  • Shows antidepressant-like effect
  • Reduces the risk of neonatal neural tube defects (sodium form)
  • Used for research of megaloblastic and macrocytic anemias due to folic deficiency