Learn More
Medchemexpress LLC Folic acid (Vitamin B9) | 59-30-3 | 99.0% | 441.40 | 50 G
SDP
Catalog No. 5000392462
Encompass_Preferred
Folic acid (Vitamin B9) is an orally active essential nutrient from the B complex group of vitamins. It exhibits an antidepressant-like effect and its sodium form reduces the risk of neonatal neural tube defects. It is used in the research of megaloblastic and macrocytic anemias due to folic deficiency.
  • Orally active essential nutrient from the B complex group of vitamins.
  • Shows antidepressant-like effect.
  • Sodium form reduces the risk of neonatal neural tube defects.
  • Can be used for research on megaloblastic and macrocytic anemias due to folic deficiency.
  • Plays a critical role in the prevention of chromosome breakage and hypomethylation of DNA.
Catalog No. 50-003-92462 Supplier Medchemexpress LLC Supplier No. HY1663750G
Folic acid (Vitamin B9) is an orally active essential nutrient from the B complex group of vitamins. It exhibits an antidepressant-like effect and its sodium form reduces the risk of neonatal neural tube defects. It is used in the research of megaloblastic and macrocytic anemias due to folic deficiency.
  • Orally active essential nutrient from the B complex group of vitamins.
  • Shows antidepressant-like effect.
  • Sodium form reduces the risk of neonatal neural tube defects.
  • Can be used for research on megaloblastic and macrocytic anemias due to folic deficiency.
  • Plays a critical role in the prevention of chromosome breakage and hypomethylation of DNA.