Learn More
Medchemexpress LLC HY-108703A 10mg Medchemexpress, Foliglurax (monohydrochloride) CAS:2133294-96-7 Purity:>98%
SDP
Catalog No. 501968615
Encompass_Preferred
Medchemexpress, HY-108703A 10mg Foliglurax (monoHCl) CAS:2133294-96-7 Foliglurax monohydrochloride (PXT002331 monohydrochloride) is a highly selective and potent, brain-penetrant metabotropic glutamate receptor 4 positive allosteric modulator (mGluR4 PAM) , with an EC50 of 79 nM. Antiparkinsonian effect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8615 Supplier Medchemexpress LLC Supplier No. HY108703A10MG
Medchemexpress, HY-108703A 10mg Foliglurax (monoHCl) CAS:2133294-96-7 Foliglurax monohydrochloride (PXT002331 monohydrochloride) is a highly selective and potent, brain-penetrant metabotropic glutamate receptor 4 positive allosteric modulator (mGluR4 PAM) , with an EC50 of 79 nM. Antiparkinsonian effect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.