Learn More
Medchemexpress LLC Foliglurax monohydrochloride | 2133294-96-7 | 98.57% | 457.97 | 10 MG
SDP
Catalog No. 5000344653
Encompass_Preferred
Foliglurax monohydrochloride is a highly selective and potent, brain-penetrant metabotropic glutamate receptor 4 positive allosteric modulator (mGluR4 PAM), with an EC50 of 79 nM. It exhibits an antiparkinsonian effect.
  • Highly selective and potent.
  • Brain-penetrant mGluR4 PAM.
  • EC50 of 79 nM for mGlu4.
  • Exhibits antiparkinsonian effect.
Catalog No. 50-003-44653 Supplier Medchemexpress LLC Supplier No. HY108703A10MM/1ML
Foliglurax monohydrochloride is a highly selective and potent, brain-penetrant metabotropic glutamate receptor 4 positive allosteric modulator (mGluR4 PAM), with an EC50 of 79 nM. It exhibits an antiparkinsonian effect.
  • Highly selective and potent.
  • Brain-penetrant mGluR4 PAM.
  • EC50 of 79 nM for mGlu4.
  • Exhibits antiparkinsonian effect.