Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Folin and Ciocalteu's Phenol Reagent, Sigma-Aldrich

Catalog No. 01259001 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-259-001 500 mL
01-258-857 100 mL
01-258-961 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-259-001 Supplier MilliporeSigma Supelco Supplier No. F9252500ML
Add to Cart
Add to Cart

Folin and Ciocalteu's phenol reagent has been used for the estimation of total phenolic content in samples.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Folin and Ciocalteu’s Phenol Reagent
Concentration 1.9 to 2.1 N
Packaging Glass Bottle
pH <0.5
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.